2-(Pyridin-4-yl)-1,5,6,7-tetrahydro-4H-pyrrolo[3,2-c]pyridin-4-one hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: PHA-767491 hydrochloride

A669599-10MG A669599-1MG A669599-50MG A669599-5MG
77254-886EA 27.81 USD
77254-886 77254-888 77254-890 77254-892
2-(Pyridin-4-yl)-1,5,6,7-tetrahydro-4H-pyrrolo[3,2-c]pyridin-4-one hydrochloride 98%
2-(Pyridin-4-yl)-1,5,6,7-tetrahydro-4H-pyrrolo[3,2-c]pyridin-4-one hydrochloride

Precaución: for Research Use Only

Formula: C₁₂H₁₂ClN₃O
MW: 249.7 g/mol
MDL Number: MFCD11519962
CAS Number: 942425-68-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR