Optovin 98%

Supplier: Ambeed
Warning

Synonyms: 5-((2,5-Dimethyl-1-(pyridin-3-yl)-1H-pyrrol-3-yl)methylene)-2-thioxothiazolidin-4-one, (5E)-5-[(2,5-dimethyl-1-pyridin-3-ylpyrrol-3-yl)methylidene]-2-sulphanylidene-1,3-thiazolidin-4-one

A669429-10MG A669429-1MG A669429-25MG A669429-50MG A669429-5MG
77243-222EA 192.61 USD
77243-222 77243-224 77243-226 77243-228 77243-230
Optovin 98%
Optovin

Precaución: for Research Use Only

Formula: C₁₅H₁₃N₃OS₂
MW: 315.409 g/mol
CAS Number: 348575-88-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR