(S,S)-2,2'-Bis[(R)-(N,N-dimethylamino)(phenyl)methyl]-1,1'-bis(diphenylphosphino)ferrocene 98%

Supplier: Ambeed
Warning

Synonyms: (S,S)-(-)-2,2'-Bis[(R)-(N,N-dimethylamino)(phenyl)methyl]-1,1'-bis(diphenylphosphino)ferrocene

A669393-100MG A669393-1G A669393-250MG A669393-50MG
77254-878EA 61 USD
77254-878 77254-880 77254-882 77254-884
(S,S)-2,2'-Bis[(R)-(N,N-dimethylamino)(phenyl)methyl]-1,1'-bis(diphenylphosphino)ferrocene 98%
(S,S)-2,2'-Bis[(R)-(N,N-dimethylamino)(phenyl)methyl]-1,1'-bis(diphenylphosphino)ferrocene

Precaución: for Research Use Only

Formula: C₅₂H₅₀FeN₂P₂
MW: 820.76 g/mol
MDL Number: MFCD04117705
CAS Number: 210842-74-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR