JNJ-17203212 98%

Supplier: Ambeed
Danger

Synonyms: 4-(3-(Trifluoromethyl)pyridin-2-yl)-N-(5-(trifluoromethyl)pyridin-2-yl)piperazine-1-carboxamide

A668990-10MG A668990-1MG A668990-25MG A668990-50MG A668990-5MG
77243-212EA 69 USD
77243-212 77243-214 77243-216 77243-218 77243-220
JNJ-17203212 98%
JNJ-17203212

Precaución: for Research Use Only

Formula: C₁₇H₁₅F₆N₅O
MDL Number: MFCD12828764
CAS Number: 821768-06-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR