3,4-Difluoroaniline 98%

Supplier: Ambeed
Danger

A668945-10G A668945-25G
77280-630EA 10 USD
77280-630 77280-632
3,4-Difluoroaniline 98%
3,4-Difluoroaniline

Precaución: for Research Use Only

Formula: C₆H₅F₂N
MDL Number: MFCD00007761
CAS Number: 3863-11-4
UN: 2941
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR