1-Iodohexane 97% stabilized

Supplier: Ambeed
Danger

Synonyms: 1-Hexyl iodide

Stabilization: Stabilized with Copper chip

A668611-25G A668611-5G
77243-202EA 13.39 USD
77243-202 77243-204
1-Iodohexane 97% stabilized
1-Iodohexane

Precaución: for Research Use Only

Formula: C₆H₁₃I
MW: 212.07 g/mol
MDL Number: MFCD00001102
CAS Number: 638-45-9
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR