1-(3-(Chloromethyl)phenyl)ethanone 98%

Supplier: Ambeed
Danger

Synonyms: 3'-Chloromethylacetophenone

A668452-1G A668452-25G A668452-5G
77280-624EA 32.96 USD
77280-624 77280-626 77280-628
1-(3-(Chloromethyl)phenyl)ethanone 98%
1-(3-(Chloromethyl)phenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₉ClO
MW: 168.6202 g/mol
MDL Number: MFCD06658170
CAS Number: 41908-12-7
UN: 1760
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR