3-Bromo-5-ethoxy-4-methoxybenzonitrile 97%

Supplier: Ambeed
Danger

A668278-1G A668278-250MG
77280-620EA 23 USD
77280-620 77280-622
3-Bromo-5-ethoxy-4-methoxybenzonitrile 97%
3-Bromo-5-ethoxy-4-methoxybenzonitrile

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrNO₂
MDL Number: MFCD02256583
CAS Number: 515831-52-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR