1H,1H,10H,10H-Hexadecafluoro-1,10-decandiol 98%

Supplier: Ambeed
Warning

Synonyms: 1H,1H,10H,10H-Perfluoro-1,10-decandiol, 1H,1h,10h,10h-perfluoro-1,10-decanediol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-Hexadecafluoro-1,10-decandiol, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9-Hexadecafluoro-1,10-decanediol, 1H,1H,10H,10H-Perfluoro-1,10-decanediol, 1H,1H,10H,10H-Perfluorodecane-1,10-diol

A668090-25G A668090-5G
77243-198EA 200 USD
77243-198 77243-200
1H,1H,10H,10H-Hexadecafluoro-1,10-decandiol 98%
1H,1H,10H,10H-Hexadecafluoro-1,10-decandiol

Precaución: for Research Use Only

Formula: C₁₀H₆F₁₆O₂
MDL Number: MFCD00192200
CAS Number: 754-96-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR