5'-(3,5-Dicarboxyphenyl)-[1,1':3',1''-terphenyl]-3,3'',5,5''-tetracarboxylic acid 98%
Supplier: Ambeed
|
Warning
|
A667998-100MG
A667998-1G
A667998-250MG
A667998-5G
77254-852EA
16
USD
77254-852
77254-854
77254-856
77254-858
5'-(3,5-Dicarboxyphenyl)-[1,1':3',1''-terphenyl]-3,3'',5,5''-tetracarboxylic acid 98%
5'-(3,5-Dicarboxyphenyl)-[1,1':3',1''-terphenyl]-3,3'',5,5''-tetracarboxylic acid
Precaución: for Research Use Only
Formula:
C₃₀H₁₈O₁₂ |
MDL Number:
MFCD30541554 CAS Number: 1228047-99-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...