3-Bromo-6-methyl-2-(trifluoromethyl)pyridine 97%

Supplier: Ambeed
Warning

A667783-100MG A667783-1G A667783-250MG
77243-192EA 25 USD
77243-192 77243-194 77243-196
3-Bromo-6-methyl-2-(trifluoromethyl)pyridine 97%
3-Bromo-6-methyl-2-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₇H₅BrF₃N
MW: 240.02 g/mol
MDL Number: MFCD18255957
CAS Number: 1211517-98-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR