2-(2-Nitrovinyl)thiophene 97%

Supplier: Ambeed
Warning

Synonyms: 2-(2-Nitroethenyl)thiophene

A667509-1G A667509-5G
77243-188EA 33.99 USD
77243-188 77243-190
2-(2-Nitrovinyl)thiophene 97%
2-(2-Nitrovinyl)thiophene

Precaución: for Research Use Only

Formula: C₆H₅NO₂S
MW: 155.17 g/mol
MDL Number: MFCD00069434
CAS Number: 874-84-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR