Ethyl 2-(tetrahydro-2H-thiopyran-4-yl)acetate 97%

Supplier: Ambeed
Warning

A667142-100MG A667142-250MG
77254-842EA 121.54 USD
77254-842 77254-844
Ethyl 2-(tetrahydro-2H-thiopyran-4-yl)acetate 97%
Ethyl 2-(tetrahydro-2H-thiopyran-4-yl)acetate

Precaución: for Research Use Only

Formula: C₉H₁₆O₂S
MW: 188.28 g/mol
MDL Number: MFCD19688077
CAS Number: 218624-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR