4-[2-Hydroxyethyl(methyl)amino]benzeneboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 2-{Methyl-[4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-benzyl]-amino}-ethanol, 2-(Methyl(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)amino)ethanol

A667096-1G A667096-5G
77243-178EA 98 USD
77243-178 77243-180
4-[2-Hydroxyethyl(methyl)amino]benzeneboronic acid pinacol ester 97%
4-[2-Hydroxyethyl(methyl)amino]benzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₆H₂₆BNO₃
MW: 291.20 g/mol
MDL Number: MFCD16294544
CAS Number: 1206641-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR