5,6-Difluoroindole-2-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5,6-Difluoro-1H-indole-2-carboxylic acid

A666413-1G A666413-250MG A666413-5G
77280-610EA 49.44 USD
77280-610 77280-612 77280-614
5,6-Difluoroindole-2-carboxylic acid 98%
5,6-Difluoroindole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₅F₂NO₂
MW: 197.138 g/mol
MDL Number: MFCD01863160
CAS Number: 169674-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR