6-Methyl-5-(trifluoromethyl)pyridin-3-amine 97%

Supplier: Ambeed
Warning

A666198-100MG A666198-250MG A666198-5G A666198-1G
77267-252EA 51 USD
77267-252 77267-254 77267-256 77749-370
6-Methyl-5-(trifluoromethyl)pyridin-3-amine 97%
6-Methyl-5-(trifluoromethyl)pyridin-3-amine

Precaución: for Research Use Only

Formula: C₇H₇F₃N₂
MDL Number: MFCD20923727
CAS Number: 1402664-67-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR