2-(2-Bromoethyl)-1,3-dioxane 98%

Supplier: Ambeed
Warning

A666007-100G A666007-10G A666007-1G A666007-25G A666007-5G
77243-162EA 175.1 USD
77243-162 77243-164 77243-166 77243-168 77243-170
2-(2-Bromoethyl)-1,3-dioxane 98%
2-(2-Bromoethyl)-1,3-dioxane

Precaución: for Research Use Only

Formula: C₆H₁₁BrO₂
MW: 195.06 g/mol
MDL Number: MFCD00006567
CAS Number: 33884-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR