2,3-Dichloro-6-methylpyridine 98%

Supplier: Ambeed
Warning

A665944-10G A665944-1G A665944-250MG A665944-25G A665944-5G
77254-832EA 130 USD
77254-832 77254-834 77254-836 77254-838 77254-840
2,3-Dichloro-6-methylpyridine 98%
2,3-Dichloro-6-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅Cl₂N
MDL Number: MFCD09839232
CAS Number: 54957-86-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR