1-Dodecanesulfonyl chloride 72% + (by ELSD)

Supplier: Ambeed
Danger

A664920-1G A664920-25G A664920-5G
77243-150EA 19 USD
77243-150 77243-152 77243-154
1-Dodecanesulfonyl chloride 72% + (by ELSD)
1-Dodecanesulphonyl chloride

Precaución: for Research Use Only

Formula: C₁₂H₂₅ClO₂S
MDL Number: MFCD00014728
CAS Number: 10147-40-7
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR