2,5-Dihydro-4-methyl-2,5-dioxo-3-furanpropanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-(4-Methyl-2,5-dioxo-2,5-dihydrofuran-3-yl)propanoic acid, 2-(2-Carboxyethyl)-3-methylmaleic Anhydride

A664682-1G A664682-250MG A664682-5G
77254-824EA 139 USD
77254-824 77254-826 77573-144
2,5-Dihydro-4-methyl-2,5-dioxo-3-furanpropanoic acid 95%
2,5-Dihydro-4-methyl-2,5-dioxo-3-furanpropanoic acid

Precaución: for Research Use Only

Formula: C₈H₈O₅
MW: 184.15 g/mol
MDL Number: MFCD18252872
CAS Number: 487-66-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR