Dimethyl 2-(4-bromophenyl)malonate 98%

Supplier: Ambeed
Warning

A664265-1G A664265-250MG A664265-25G A664265-5G
77243-140EA 106.09 USD
77243-140 77243-142 77243-144 77243-146
Dimethyl 2-(4-bromophenyl)malonate 98%
Dimethyl 2-(4-bromophenyl)malonate

Precaución: for Research Use Only

Formula: C₁₁H₁₁BrO₄
MW: 287.10 g/mol
MDL Number: MFCD06204131
CAS Number: 149506-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR