Dde-Dap(Fmoc)-OH 98%

Supplier: Ambeed
Warning

A663862-1G A663862-250MG A663862-5G
77243-130EA 110.21 USD
77243-130 77243-132 77243-134
Dde-Dap(Fmoc)-OH 98%
Dde-Dap(Fmoc)-OH

Precaución: for Research Use Only

Formula: C₂₈H₃₀N₂O₆
MW: 490.54 g/mol
MDL Number: MFCD05663745
CAS Number: 1263046-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR