Methyl 2,3,4-Tri-O-acetyl-b-D-glucuronic Acid Methyl Ester 98%

Supplier: Ambeed
Warning

A663419-1G A663419-5G
77243-122EA 8 USD
77243-122 77243-124
Methyl 2,3,4-Tri-O-acetyl-b-D-glucuronic Acid Methyl Ester 98%
Methyl 2,3,4-Tri-O-acetyl-b-D-glucuronic Acid Methyl Ester

Precaución: for Research Use Only

Formula: C₁₄H₂₀O₁₀
MDL Number: MFCD09841789
CAS Number: 34213-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR