1,1′-Bis[(2R,5R)-2,5-diethylphospholano]ferrocene ≥98%, ee 99%

Supplier: Ambeed
Warning

A662878-100MG A662878-10G A662878-1G A662878-250MG A662878-5G
77254-806EA 31 USD
77254-806 77254-808 77254-810 77254-812 77254-814
1,1′-Bis[(2R,5R)-2,5-diethylphospholano]ferrocene ≥98%, ee 99%
1,1′-Bis[(2R,5R)-2,5-diethylphospholano]ferrocene

Precaución: for Research Use Only

Formula: C₂₆H₄₀FeP₂
MW: 470.38 g/mol
MDL Number: MFCD09265149
CAS Number: 147762-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR