3-(Naphthalen-2-yl)benzaldehyde 97%

Supplier: Ambeed
Warning

A662729-100MG A662729-1G A662729-250MG
77254-800EA 19 USD
77254-800 77254-802 77254-804
3-(Naphthalen-2-yl)benzaldehyde 97%
3-(Naphthalen-2-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₇H₁₂O
MDL Number: MFCD04117446
CAS Number: 728919-26-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR