[1,1':4',1''-Terphenyl]-2',3,3'',5,5',5''-hexacarboxylic acid 98%

Supplier: Ambeed
Warning

A662475-100MG A662475-250MG A662475-1G
77267-238EA 24 USD
77267-238 77267-240 77749-363
[1,1':4',1''-Terphenyl]-2',3,3'',5,5',5''-hexacarboxylic acid 98%
[1,1':4',1''-Terphenyl]-2',3,3'',5,5',5''-hexacarboxylic acid

Precaución: for Research Use Only

Formula: C₂₄H₁₄O₁₂
CAS Number: 1542274-12-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR