3,5-Difluoro-4-methoxybenzyl alcohol 95%

Supplier: Ambeed
Warning

Synonyms: 2,6-Difluoro-4-(hydroxymethyl)anisole, 3,5-Difluoro-4-methoxybenzyl Alcohol

A662402-250MG A662402-5G
77280-582EA 18.54 USD
77280-582 77280-584
3,5-Difluoro-4-methoxybenzyl alcohol 95%
3,5-Difluoro-4-methoxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₈F₂O₂
MW: 174.145 g/mol
MDL Number: MFCD04115914
CAS Number: 706786-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR