NSC-59984 98%

Supplier: Ambeed
Warning

Synonyms: NSC59984, (E)-1-(4-methylpiperazin-1-yl)-3-(5-nitrofuran-2-yl)prop-2-en-1-one

A661348-10MG A661348-1MG A661348-25MG A661348-2MG A661348-50MG A661348-5MG
77243-098EA 19 USD
77243-098 77243-100 77243-102 77243-104 77243-106 77243-108
NSC-59984 98%
NSC-59984

Precaución: for Research Use Only

Formula: C₁₂H₅N₃O₄
MDL Number: MFCD29905462
CAS Number: 803647-40-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR