2-Methylthiophene 98%

Supplier: Ambeed
Danger

A661002-10G A661002-25G A661002-5G
77280-568EA 7 USD
77280-568 77280-570 77280-572
2-Methylthiophene 98%
2-Methylthiophene

Precaución: for Research Use Only

Formula: C₅H₆S
MDL Number: MFCD00005451
CAS Number: 554-14-3
UN: 1993
ADR: 3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR