2,5-Dimercaptoterephthalic acid 98% (by HPLC)

Supplier: Ambeed
Warning

A660942-100MG A660942-250MG A660942-5G A660942-1G
77267-222EA 16 USD
77267-222 77267-224 77267-226 77617-762
2,5-Dimercaptoterephthalic acid 98% (by HPLC)
2,5-Dimercaptoterephthalic acid

Precaución: for Research Use Only

Formula: C₈H₆O₄S₂
MDL Number: MFCD23160366
CAS Number: 25906-66-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR