N-(3-(1H-Imidazol-1-yl)propyl)-5-(furan-2-yl)isoxazole-3-carboxamide 98%

Supplier: Ambeed
Warning

Synonyms: SKL2001

A660516-100MG A660516-10MG A660516-1MG A660516-50MG A660516-5MG
77267-210EA 130.81 USD
77267-210 77267-212 77267-214 77267-216 77267-218
N-(3-(1H-Imidazol-1-yl)propyl)-5-(furan-2-yl)isoxazole-3-carboxamide 98%
N-(3-(1H-Imidazol-1-yl)propyl)-5-(furan-2-yl)isoxazole-3-carboxamide

Precaución: for Research Use Only

Formula: C₁₄H₁₄N₄O₃
MW: 286.29 g/mol
MDL Number: MFCD14733081
CAS Number: 909089-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR