3-Bromo-2-chloro-6-(difluoromethyl)pyridine 97%

Supplier: Ambeed
Warning

A660455-100MG A660455-250MG A660455-50MG
77243-092EA 251.32 USD
77243-092 77243-094 77243-096
3-Bromo-2-chloro-6-(difluoromethyl)pyridine 97%
3-Bromo-2-chloro-6-(difluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₃BrClF₂N
MW: 242.44 g/mol
MDL Number: MFCD25477647
CAS Number: 1805221-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR