(4-Chlorophenoxy)acetonitrile 97%

Supplier: Ambeed
Danger

A660133-10G A660133-25G
77243-088EA 13 USD
77243-088 77243-090
(4-Chlorophenoxy)acetonitrile 97%
(4-Chlorophenoxy)acetonitrile

Precaución: for Research Use Only

Formula: C₈H₆ClNO
MDL Number: MFCD00031535
CAS Number: 3598-13-8
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR