1,1'-Bis[(2S,5S)-2,5-dimethyl-1-phospholanyl]ferrocene 97%

Supplier: Ambeed
Warning

Synonyms: 1,1-Bis((2S,5S)-2,5-dimethylphospholano)ferrocene

A659763-100MG A659763-10G A659763-1G A659763-250MG A659763-5G
77254-784EA 21 USD
77254-784 77254-786 77254-788 77254-790 77254-792
1,1'-Bis[(2S,5S)-2,5-dimethyl-1-phospholanyl]ferrocene 97%
1,1'-Bis[(2S,5S)-2,5-dimethyl-1-phospholanyl]ferrocene

Precaución: for Research Use Only

Formula: C₂₂H₃₂FeP₂
MW: 414.291 g/mol
MDL Number: MFCD09265089
CAS Number: 162412-87-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR