N,N'-[(1R,2R)-1,2-Diphenyl-1,2-ethanediyl]bis[2-diphenylphosphinobenzamide] ≥97%, ee 99%

Supplier: Ambeed
Warning

A659556-100MG A659556-1G
77254-780EA 26.78 USD
77254-780 77254-782
N,N'-[(1R,2R)-1,2-Diphenyl-1,2-ethanediyl]bis[2-diphenylphosphinobenzamide] ≥97%, ee 99%
N,N'-[(1R,2R)-1,2-Diphenyl-1,2-ethanediyl]bis[2-diphenylphosphinobenzamide]

Precaución: for Research Use Only

Formula: C₅₂H₄₂N₂O₂P₂
MW: 788.84 g/mol
MDL Number: MFCD30541655
CAS Number: 138517-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR