2-(Chloromethyl)quinoline 98%

Supplier: Ambeed
Danger

A659498-1G A659498-5G
77243-084EA 98.88 USD
77243-084 77243-086
2-(Chloromethyl)quinoline 98%
2-(Chloromethyl)quinoline

Precaución: for Research Use Only

Formula: C₁₀H₈ClN
MW: 177.63 g/mol
MDL Number: MFCD00023999
CAS Number: 4377-41-7
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR