3-Chloro-5-iodobenzonitrile 98%

Supplier: Ambeed
Warning

A659450-100G A659450-1G A659450-25G A659450-5G
77280-560EA 748.81 USD
77280-560 77280-562 77280-564 77280-566
3-Chloro-5-iodobenzonitrile 98%
3-Chloro-5-iodobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃ClIN
MW: 263.4629 g/mol
MDL Number: MFCD00672974
CAS Number: 289039-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR