4,4'-Diphenylbenzophenone 98%

Supplier: Ambeed
Warning

Synonyms: Di([1,1'-biphenyl]-4-yl)methanone

A659402-1G A659402-250MG A659402-25G A659402-5G
77293-090EA 27.81 USD
77293-090 77293-092 77293-094 77293-096
4,4'-Diphenylbenzophenone 98%
4,4'-Diphenylbenzophenone

Precaución: for Research Use Only

Formula: C₂₅H₁₈O
MW: 334.42 g/mol
MDL Number: MFCD00016575
CAS Number: 3478-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR