Ethyl 4-amino-2-hydroxypyrimidine-5-carboxylate 97%

Supplier: Ambeed
Warning

Synonyms: 5-Carbethoxycytosine, Ethyl 4-amino-2-hydroxypyrimidine-5-carboxylate, 4-Amino-2-hydroxy-pyrimidine-5-carboxylic acid ethyl ester

A658907-100MG A658907-1G
77293-088EA 77.25 USD
77293-088 77617-754
Ethyl 4-amino-2-hydroxypyrimidine-5-carboxylate 97%
Ethyl 4-amino-2-hydroxypyrimidine-5-carboxylate

Precaución: for Research Use Only

Formula: C₇H₉N₃O₃
MW: 183.17 g/mol
MDL Number: MFCD00552904
CAS Number: 20187-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR