(2R,3R)-1,4-dioxaspiro[4.5]decane-2,3-diylbis(diphenylmethanol) ≥98%, ee 99%

Supplier: Ambeed
Warning

A658291-100MG A658291-1G A658291-250MG A658291-5G
77254-764EA 13.39 USD
77254-764 77254-766 77254-768 77254-770
(2R,3R)-1,4-dioxaspiro[4.5]decane-2,3-diylbis(diphenylmethanol) ≥98%, ee 99%
(2R,3R)-1,4-dioxaspiro[4.5]decane-2,3-diylbis(diphenylmethanol)

Precaución: for Research Use Only

Formula: C₃₄H₃₄O₄
MW: 506.63 g/mol
MDL Number: MFCD29905016
CAS Number: 114026-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR