1-{[4-(Tetramethyl-1,3,2-dioxaborolan-2-yl)benzene]sulfonyl}piperidine 97%

Supplier: Ambeed
Warning

Synonyms: 1-((4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)sulphonyl)piperidine

A658189-1G A658189-250MG A658189-5G
77243-078EA 60.77 USD
77243-078 77243-080 77243-082
1-{[4-(Tetramethyl-1,3,2-dioxaborolan-2-yl)benzene]sulfonyl}piperidine 97%
1-{[4-(Tetramethyl-1,3,2-dioxaborolan-2-yl)benzene]sulfonyl}piperidine

Precaución: for Research Use Only

Formula: C₁₇H₂₆BNO₄S
MW: 351.269 g/mol
MDL Number: MFCD18915049
CAS Number: 1548827-83-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR