4-Fluoro-3-(trifluoromethyl)benzenesulfonyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 4-Fluoro-3-(trifluoromethyl)benzene-1-sulphonyl chloride

A657230-1G A657230-25G A657230-5G
77293-076EA 12.36 USD
77293-076 77293-078 77293-080
4-Fluoro-3-(trifluoromethyl)benzenesulfonyl chloride 98%
4-Fluoro-3-(trifluoromethyl)benzenesulphonyl chloride

Precaución: for Research Use Only

Formula: C₇H₃ClF₄O₂S
MW: 262.61 g/mol
MDL Number: MFCD01091001
CAS Number: 1682-10-6
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR