4-(Phenylethynyl)benzoic acid 97%

Supplier: Ambeed
Warning

A657197-100MG A657197-250MG A657197-1G A657197-5G
77293-072EA 13.39 USD
77293-072 77293-074 77617-752 77749-354
4-(Phenylethynyl)benzoic acid 97%
4-(Phenylethynyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₀O₂
MW: 222.25 g/mol
MDL Number: MFCD00438677
CAS Number: 25739-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR