2-Isopropyl-6-methoxypyridine 95%

Supplier: Ambeed
Warning

A656607-1G A656607-250MG A656607-5G
77267-202EA 113 USD
77267-202 77267-204 77267-206
2-Isopropyl-6-methoxypyridine 95%
2-Isopropyl-6-methoxypyridine

Precaución: for Research Use Only

Formula: C₉H₁₃NO
MDL Number: MFCD09027948
CAS Number: 479412-25-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR