2,3-Dichloro-5-(trichloromethyl)pyridine 95%

Supplier: Ambeed
Warning

A656516-10G A656516-250MG
77243-074EA 648.99 USD
77243-074 77243-076
2,3-Dichloro-5-(trichloromethyl)pyridine 95%
2,3-Dichloro-5-(trichloromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₂Cl₅N
MDL Number: MFCD03701543
CAS Number: 69045-83-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR