4-Iodo-3-nitroaniline 97%

Supplier: Ambeed
Warning

A655925-1G A655925-25G
77254-736EA 24.72 USD
77254-736 77254-738
4-Iodo-3-nitroaniline 97%
4-Iodo-3-nitroaniline

Precaución: for Research Use Only

Formula: C₆H₅IN₂O₂
MW: 264.02 g/mol
MDL Number: MFCD08458843
CAS Number: 105752-04-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR