Methyl 2-(3-formyl-4-hydroxyphenyl)acetate 98%

Supplier: Ambeed
Warning

A655804-1G A655804-5G A655804-250MG
77293-050EA 284.28 USD
77293-050 77293-052 77617-746
Methyl 2-(3-formyl-4-hydroxyphenyl)acetate 98%
Methyl 2-(3-formyl-4-hydroxyphenyl)acetate

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
MW: 194.18 g/mol
MDL Number: MFCD04972464
CAS Number: 61874-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR