4-Benzyloxy-3-fluorophenylboronic acid 98%

Supplier: Ambeed
Warning

A655201-1G A655201-25G A655201-5G
77280-546EA 10 USD
77280-546 77280-548 77280-550
4-Benzyloxy-3-fluorophenylboronic acid 98%
4-Benzyloxy-3-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₂BFO₃
MDL Number: MFCD00994627
CAS Number: 133057-83-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR