[2,2'-Bipyridin]-4-amine 95%

Supplier: Ambeed
Warning

A654946-1G A654946-250MG A654946-100MG
77254-732EA 186 USD
77254-732 77254-734 77617-744
[2,2'-Bipyridin]-4-amine 95%
[2,2'-Bipyridin]-4-amine

Precaución: for Research Use Only

Formula: C₁₀H₉N₃
MDL Number: MFCD01646268
CAS Number: 14151-21-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR