2-(Bromomethyl)-2-(hydroxymethyl)-1,3-propanediol 98%

Supplier: Ambeed
Warning

Synonyms: 2-(Bromomethyl)-2-(hydroxymethyl)propane-1,3-diol

A654856-1G A654856-10G
77243-072EA 27.81 USD
77243-072 77551-988
2-(Bromomethyl)-2-(hydroxymethyl)-1,3-propanediol 98%
2-(Bromomethyl)-2-(hydroxymethyl)-1,3-propanediol

Precaución: for Research Use Only

Formula: C₅H₁₁BrO₃
MW: 199.04 g/mol
MDL Number: MFCD00135306
CAS Number: 19184-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR